Learn More
Waters Corp Waters Corporation Method Validation Kit, C8, 2.5 μm, 3 mm × 150 mm, 3/pk
Supplier:  Waters Corp 186006761
Encompass_Preferred
This method validation kit includes three columns, each packed with a different batch of sorbent. The C8 sorbent offers less retention compared to C18 sorbent, which can be advantageous for faster method development. It also helps improve peak shape and chromatographic performance, facilitating a seamless transition to UPLC.
  • Trifunctionally bonded sorbent provides the widest usable pH range (1–12).
  • Superior low pH stability.
  • Ultra–low column bleed.
  • Packed in ultra–low dispersion hardware.
  • 2.5 μm eXtended Performance [XP] columns enable exceptional separation performance, robustness, and throughput for HPLC assays.
Catalog No. 50-466-463