Learn More
Waters Corp XBridge BEH C8 Method Validation Kit, 130Å, 2.5 µm, 4.6 mm X 150 mm, 3/pk
Catalog No. 50466464 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This method validation kit consists of three XBridge BEH C8 columns packed with three different batches of sorbent. The XBridge BEH C8 sorbent is less retentive compared to the XBridge BEH C18 sorbent. Anyone developing methods may prefer the speed offered by a less retentive column, while improving peak shape and chromatographic performance. The trifunctionally bonded sorbent offers you the widest usable pH range (1-12), superior low pH stability, and ultra-low column bleed. Packed in ultra-low dispersion hardware, 2.5 µm eXtended Performance [XP] columns enable exceptional separation performance, robustness and throughput for HPLC assays while enabling a seamless transition path toward UPLC
Catalog No. 50-466-464 Supplier Waters Corp Supplier No. 186006762
This method validation kit consists of three XBridge BEH C8 columns packed with three different batches of sorbent. The XBridge BEH C8 sorbent is less retentive compared to the XBridge BEH C18 sorbent. Anyone developing methods may prefer the speed offered by a less retentive column, while improving peak shape and chromatographic performance. The trifunctionally bonded sorbent offers you the widest usable pH range (1-12), superior low pH stability, and ultra-low column bleed. Packed in ultra-low dispersion hardware, 2.5 µm eXtended Performance [XP] columns enable exceptional separation performance, robustness and throughput for HPLC assays while enabling a seamless transition path toward UPLC