Learn More
Waters Corp Waters Column, C8, 130Å, 2.5 μm, 4.6 mm × 150 mm
Supplier:  Waters Corp 186006714
Encompass_Preferred
The XBridge BEH C8 column features a less retentive sorbent than C18, enabling faster methods and improved peak shape. It offers a wide usable pH range (1-12) and superior low pH stability, along with ultra-low column bleed. The 2.5 μm XP columns, packed in ultra-low dispersion hardware, provide exceptional separation performance for HPLC assays and a seamless transition to UPLC.
  • Less retentive than C18 sorbent for faster methods and improved peak shape.
  • Features a wide usable pH range (1-12), superior low pH stability, and ultra-low column bleed.
  • Provides exceptional separation performance for HPLC assays and enables a seamless transition to UPLC.
  • Designed for increased assay robustness, a longer column lifespan, and unrivaled reproducibility.
Catalog No. 50-466-422