Learn More
Waters Corp WATERS CORPORATION Direct Connect HP Column, C8, 10 μm, 2.1 mm × 30 mm
Catalog No. 502037693 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The XBridge C8 Direct Connect HP 2.1×30 mm column is engineered for automated UPLC sample preparation. It delivers excellent peak shape, high efficiency, stability across acidic and basic mobile phases, and supports an operating pH range of 1 to 12. It integrates with ACQUITY UPLC for analyzing specific analytes.
  • Engineered for automated sample preparation with UPLC On-line SPE Technology.
  • Provides excellent peak shape, high efficiency, and stability for acidic/basic mobile phases.
  • Features UPLC-rated direct connect hardware for convenient column integration.
  • Ensures improved reliability, assay robustness, and extended column lifetime.
  • Maximizes particle efficiency for unrivaled peak form, capacity, and reproducibility.
  • cGMP, ISO 9001-certified manufacturing with chromatographically tested batches.
Catalog No. 50-203-7693 Supplier Waters Corp Supplier No. 186005233
The XBridge C8 Direct Connect HP 2.1×30 mm column is engineered for automated UPLC sample preparation. It delivers excellent peak shape, high efficiency, stability across acidic and basic mobile phases, and supports an operating pH range of 1 to 12. It integrates with ACQUITY UPLC for analyzing specific analytes.
  • Engineered for automated sample preparation with UPLC On-line SPE Technology.
  • Provides excellent peak shape, high efficiency, and stability for acidic/basic mobile phases.
  • Features UPLC-rated direct connect hardware for convenient column integration.
  • Ensures improved reliability, assay robustness, and extended column lifetime.
  • Maximizes particle efficiency for unrivaled peak form, capacity, and reproducibility.
  • cGMP, ISO 9001-certified manufacturing with chromatographically tested batches.