Learn More
Waters Corp Waters Corporation OBD Prep Column, BEH C8, 10 μm, 10 x 150 mm
Supplier:  Waters Corp 186008215
Encompass_Preferred
Waters XBridge Preparative Columns with BEH C8 chemistry provide exceptional performance for diverse compounds, offering superior peak shape for basic analytes at high pH and stability across broad pH ranges and temperatures. Their C1-C8 alkyl chain suits strongly hydrophobic compounds, ensuring enhanced Prep LC via uniform packed beds, direct scalability, and consistent, long-lasting results.
  • Universal column for diverse compounds with BEH Technology™.
  • Exceptional peak shape for basic analytes at high pH.
  • Stable across wide pH and high temperatures.
  • Shorter C1-C8 alkyl chain ideal for strongly hydrophobic compounds.
  • Uniform packed bed density for enhanced Prep LC.
  • Direct analytical to preparative scalability.
  • Excellent column lifetime.
  • Consistent column-to-column performance.
  • Ensures superior resolution, peak shape, and mass loading.
Catalog No. 50-239-2828