Learn More
Waters Corp Waters XBridge BEH C8 OBD Prep Column, 10 μm, 10 x 250 mm
Supplier:  Waters Corp 186008216
Encompass_Preferred
This OBD Prep Column is designed for high-speed applications, offering less retention than C18 sorbents. It features a trifunctionally bonded sorbent for a wide usable pH range (1 to 12), superior low pH stability, and ultra-low column bleed, enhancing peak shape and chromatographic performance.
  • Less retentive than C18 sorbents, ideal for high-speed methods.
  • Widest usable pH range (1 to 12).
  • Superior low pH stability and ultra-low column bleed.
  • Improves peak shape and chromatographic performance.
  • Ensures reproducible column performance through extensive testing.
  • Offers increased pH stability and longer column life.
  • Delivers higher assay robustness and reliability.
  • Provides unmatched peak form and capacity, maximizing particle efficiency.
  • Features high efficiency, symmetrical peak form, and outstanding pH stability.
Catalog No. 50-203-8419