Learn More
Waters Corp VanGuard Cartridge, C8, 5 μm, 130 Å, 3.9 x 5 mm
Supplier: Waters Corp 186007780
VanGuard Cartridges extend the lifetime and performance of analytical columns by removing particulate contamination. This cartridge is optimized for 4.6 mm I.D. analytical columns with 5 μm sorbent particles, resisting high pressures for routine use.
- Extends analytical column lifetime and performance.
- Effectively removes particulate contamination.
- Optimized for 4.6 mm I.D. analytical columns with 5 μm sorbent particles.
- Resists high pressures (up to 18,000 psi / 1240 Bar).
- Utilizes 130 Å pore, 5 μm BEH Technology C8 particles.
- Each batch undergoes quality control testing via peptide mapping.
- Designed for reversed-phase UPLC liquid chromatography.
- Easy to install and use.
- Protects columns from damaging contaminants.
- Offers a broad pH range of 1–12.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.