Learn More
Waters Corp XBridge VanGuard Cartridge, BEH C8, 130Å, 5 μm, 2.1 mm x 5 mm
Catalog No. 502391908 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
XBridge BEH C8 VanGuard Cartridges are designed to extend the lifetime and performance of analytical columns. They achieve this by removing particulate contamination from the mobile phase stream. This specific cartridge is optimized for use with all 2.1 mm and 3.0 mm I.D. XBridge BEH C8 analytical columns that contain 5 μm sorbent particles.
  • Extends analytical column lifetime and performance
  • Removes particulate contamination from the mobile phase stream
  • Optimized for 2.1 mm and 3.0 mm I.D. analytical columns with 5 μm sorbent particles
Catalog No. 50-239-1908 Supplier Waters Corp Supplier No. 186007778
XBridge BEH C8 VanGuard Cartridges are designed to extend the lifetime and performance of analytical columns. They achieve this by removing particulate contamination from the mobile phase stream. This specific cartridge is optimized for use with all 2.1 mm and 3.0 mm I.D. XBridge BEH C8 analytical columns that contain 5 μm sorbent particles.
  • Extends analytical column lifetime and performance
  • Removes particulate contamination from the mobile phase stream
  • Optimized for 2.1 mm and 3.0 mm I.D. analytical columns with 5 μm sorbent particles