Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Waters Corporation HPLC Column, XBridge BEH C8, 2.5 μm, 3 × 100 mm

Catalog No. 50179481 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

This BEH C8 sorbent is ideal for method development, delivering faster speeds and improved peak shape. It features a wide usable pH range (1 to 12), excellent low pH stability, and minimal column bleed. The 2.5 μm XP particles ensure high separation performance, robustness, and throughput for HPLC assays, with a smooth transition to UPLC.

  • C8 chemistry
  • Reversed phase separation mode
  • Particle size: 2.5 μm
  • pH range: 1 to 12
  • Maximum pressure: 18000 psi (1240 bar)
  • Pore size: 130 Å
  • Inner diameter: 3 mm
  • Length: 100 mm
  • System compatibility: UPLC, UHPLC

Catalog No. 50-179-481 Supplier Waters Corp Supplier No. 186006047
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This BEH C8 sorbent is ideal for method development, delivering faster speeds and improved peak shape. It features a wide usable pH range (1 to 12), excellent low pH stability, and minimal column bleed. The 2.5 μm XP particles ensure high separation performance, robustness, and throughput for HPLC assays, with a smooth transition to UPLC.

  • C8 chemistry
  • Reversed phase separation mode
  • Particle size: 2.5 μm
  • pH range: 1 to 12
  • Maximum pressure: 18000 psi (1240 bar)
  • Pore size: 130 Å
  • Inner diameter: 3 mm
  • Length: 100 mm
  • System compatibility: UPLC, UHPLC

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.