Learn More
Waters Corp XBridge BEH C8 XP Column, 130Å, 2.5 µm, 2.1 mm X 30 mm, 1/pk
Catalog No. 50179474 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The XBridge BEH C8 sorbent is less retentive compared to the XBridge BEH C18 sorbent. Anyone developing methods may prefer the speed offered by a less retentive column, while improving peak shape and chromatographic performance. The trifunctionally bonded sorbent offers you the widest usable pH range (1-12), superior low pH stability, and ultra-low column bleed. Packed in ultra-low dispersion hardware, 2.5 µm eXtended Performance [XP] columns enable exceptional separation performance, robustness and throughput for HPLC assays while enabling a seamless transition path toward UPLC adoption.
Catalog No. 50-179-474 Supplier Waters Corp Supplier No. 186006040
The XBridge BEH C8 sorbent is less retentive compared to the XBridge BEH C18 sorbent. Anyone developing methods may prefer the speed offered by a less retentive column, while improving peak shape and chromatographic performance. The trifunctionally bonded sorbent offers you the widest usable pH range (1-12), superior low pH stability, and ultra-low column bleed. Packed in ultra-low dispersion hardware, 2.5 µm eXtended Performance [XP] columns enable exceptional separation performance, robustness and throughput for HPLC assays while enabling a seamless transition path toward UPLC adoption.