Learn More
Waters Corp Waters Corporation Column, C8, 2.5 μm, 2.1 mm x 30 mm, 3-pack
Catalog No. 50179213 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This C8 sorbent is less retentive than C18 sorbents, offering faster analysis and improved peak shape. The trifunctionally bonded sorbent provides a wide usable pH range (1–12), superior stability at low pH, and ultra-low column bleed. When packed in ultra-low dispersion hardware, these 2.5 μm eXtended Performance (XP) columns deliver exceptional separation, robustness, and high throughput for HPLC assays, facilitating UPLC adoption.
  • Less retentive C8 sorbent for faster analysis
  • Improved peak shape and chromatographic performance
  • Trifunctionally bonded for wide pH range (1–12)
  • Superior low pH stability
  • Ultra-low column bleed
  • eXtended Performance (XP) columns deliver exceptional separation, robustness, and high throughput
  • Smooth transition to UPLC adoption
Catalog No. 50-179-213 Supplier Waters Corp Supplier No. 176002554
This C8 sorbent is less retentive than C18 sorbents, offering faster analysis and improved peak shape. The trifunctionally bonded sorbent provides a wide usable pH range (1–12), superior stability at low pH, and ultra-low column bleed. When packed in ultra-low dispersion hardware, these 2.5 μm eXtended Performance (XP) columns deliver exceptional separation, robustness, and high throughput for HPLC assays, facilitating UPLC adoption.
  • Less retentive C8 sorbent for faster analysis
  • Improved peak shape and chromatographic performance
  • Trifunctionally bonded for wide pH range (1–12)
  • Superior low pH stability
  • Ultra-low column bleed
  • eXtended Performance (XP) columns deliver exceptional separation, robustness, and high throughput
  • Smooth transition to UPLC adoption