Learn More
Waters Corp Waters XBridge BEH C8 XP Column, 130Å, 2.5 μm, 3 mm X 30 mm
Catalog No. 50179217 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This XBridge BEH C8 sorbent column offers less retention than C18, accelerating method development and improving peak shape and chromatographic performance. Its trifunctionally bonded sorbent provides a wide pH range (1 to 12), excellent low pH stability, and ultra-low column bleed. Packed in ultra-low dispersion hardware, these 2.5 μm XP columns deliver exceptional separation performance, robustness, and throughput for HPLC assays, facilitating UPLC transition.
  • Less retentive than C18 for faster method development.
  • Improved peak shape and chromatographic performance.
  • Trifunctionally bonded sorbent with a wide pH range (1 to 12).
  • Superior low pH stability.
  • Ultra-low column bleed.
  • Packed in ultra-low dispersion hardware.
  • 2.5 μm XP columns deliver exceptional separation, robustness, and throughput for HPLC.
  • Facilitates seamless UPLC transition.
Catalog No. 50-179-217 Supplier Waters Corp Supplier No. 176002558
This XBridge BEH C8 sorbent column offers less retention than C18, accelerating method development and improving peak shape and chromatographic performance. Its trifunctionally bonded sorbent provides a wide pH range (1 to 12), excellent low pH stability, and ultra-low column bleed. Packed in ultra-low dispersion hardware, these 2.5 μm XP columns deliver exceptional separation performance, robustness, and throughput for HPLC assays, facilitating UPLC transition.
  • Less retentive than C18 for faster method development.
  • Improved peak shape and chromatographic performance.
  • Trifunctionally bonded sorbent with a wide pH range (1 to 12).
  • Superior low pH stability.
  • Ultra-low column bleed.
  • Packed in ultra-low dispersion hardware.
  • 2.5 μm XP columns deliver exceptional separation, robustness, and throughput for HPLC.
  • Facilitates seamless UPLC transition.