Learn More
Waters Corp XBridge BEH C8 Method Validation Kit, 130Å, 2.5 μm, 2.1 x 100 mm
Catalog No. 50179581 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This XBridge BEH C8 Method Validation Kit is a chromatography supply designed for HPLC columns, specifically for reversed-phase chromatography applications. It includes three columns, each with a 130Å pore size, 2.5 μm particle size, and dimensions of 2.1 mm ID x 100 mm length.
  • Designed for high-performance liquid chromatography (HPLC) applications.
  • Optimized for reversed-phase chromatography.
  • Features XBridge BEH C8 phase.
  • Pore size of 130Å.
  • Particle size of 2.5 μm.
  • Column dimensions of 2.1 mm ID x 100 mm length.
  • Includes three columns for method validation.
  • Complies with USP L7 specification.
Catalog No. 50-179-581 Supplier Waters Corp Supplier No. 186006204
This XBridge BEH C8 Method Validation Kit is a chromatography supply designed for HPLC columns, specifically for reversed-phase chromatography applications. It includes three columns, each with a 130Å pore size, 2.5 μm particle size, and dimensions of 2.1 mm ID x 100 mm length.
  • Designed for high-performance liquid chromatography (HPLC) applications.
  • Optimized for reversed-phase chromatography.
  • Features XBridge BEH C8 phase.
  • Pore size of 130Å.
  • Particle size of 2.5 μm.
  • Column dimensions of 2.1 mm ID x 100 mm length.
  • Includes three columns for method validation.
  • Complies with USP L7 specification.