Learn More
Waters Corp Waters XBridge Column, C8, 130Å, 2.5 μm, 3 mm
Supplier:  Waters Corp 186006205
Encompass_Preferred
This column offers a less retentive C8 sorbent compared to C18, optimizing method development speed and improving chromatographic performance through better peak shape. It features a trifunctionally bonded sorbent for a broad usable pH range of 1-12, excellent low pH stability, and minimal column bleed. The 2.5 μm eXtended Performance (XP) columns are designed with ultra-low dispersion hardware, ensuring superior separation, robustness, and throughput for HPLC assays, while also providing a smooth upgrade path to UPLC systems.
  • Less retentive sorbent for faster method development
  • Improved peak shape and chromatographic performance
  • Widest usable pH range (1-12)
  • Superior low pH stability
  • Ultra-low column bleed
  • Exceptional separation performance and robustness
  • High throughput for HPLC assays
  • Seamless transition path to UPLC adoption
Catalog No. 50-179-582