Learn More
Waters Corp Waters XBridge BEH C8 XP Column, 2.5 μm, 2.1 × 75 mm
Catalog No. 50179215 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The C8 sorbent is less retentive than the C18 sorbent, which can be beneficial for method development due to faster speed while improving peak shape and chromatographic performance. The trifunctionally bonded sorbent offers a wide usable pH range of 1 to 12, superior low pH stability, and ultra-low column bleed.
  • Trifunctionally bonded sorbent.
  • Offers a wide usable pH range of 1 to 12.
  • Provides superior low pH stability.
  • Ensures ultra-low column bleed.
  • Packed in ultra-low dispersion hardware for exceptional separation performance.
  • Delivers robustness and high throughput for HPLC assays.
  • Enables a seamless transition towards UPLC adoption.
Catalog No. 50-179-215 Supplier Waters Corp Supplier No. 176002556
The C8 sorbent is less retentive than the C18 sorbent, which can be beneficial for method development due to faster speed while improving peak shape and chromatographic performance. The trifunctionally bonded sorbent offers a wide usable pH range of 1 to 12, superior low pH stability, and ultra-low column bleed.
  • Trifunctionally bonded sorbent.
  • Offers a wide usable pH range of 1 to 12.
  • Provides superior low pH stability.
  • Ensures ultra-low column bleed.
  • Packed in ultra-low dispersion hardware for exceptional separation performance.
  • Delivers robustness and high throughput for HPLC assays.
  • Enables a seamless transition towards UPLC adoption.