Learn More
Waters Corp Waters Column, C8, 2.5 μm, 4.6 mm x 30 mm, 130 Å
Catalog No. 50201518 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The XBridge BEH C8 sorbent is ideal for method development, offering lower retention than C18 for improved speed and chromatographic performance. This trifunctionally bonded sorbent provides a wide pH range (1 to 12), superior low pH stability, and ultra-low column bleed. These 2.5 μm XP columns deliver excellent separation, robustness, and throughput for HPLC, facilitating seamless UPLC transition.
  • Lower retention than C18 for faster method development.
  • Wide pH range (1 to 12), superior low pH stability, ultra-low column bleed.
  • Trifunctionally bonded sorbent.
  • XP columns with ultra-low dispersion hardware for exceptional separation.
  • Robustness and throughput for HPLC; seamless UPLC transition.
  • Low silanol activity, BEH particle technology, spherical shape.
  • Endcapped, Parker-style endfitting, UPLC/UHPLC compatible.
  • USP classification: L7.
Catalog No. 50-201-518 Supplier Waters Corp Supplier No. 186006048
The XBridge BEH C8 sorbent is ideal for method development, offering lower retention than C18 for improved speed and chromatographic performance. This trifunctionally bonded sorbent provides a wide pH range (1 to 12), superior low pH stability, and ultra-low column bleed. These 2.5 μm XP columns deliver excellent separation, robustness, and throughput for HPLC, facilitating seamless UPLC transition.
  • Lower retention than C18 for faster method development.
  • Wide pH range (1 to 12), superior low pH stability, ultra-low column bleed.
  • Trifunctionally bonded sorbent.
  • XP columns with ultra-low dispersion hardware for exceptional separation.
  • Robustness and throughput for HPLC; seamless UPLC transition.
  • Low silanol activity, BEH particle technology, spherical shape.
  • Endcapped, Parker-style endfitting, UPLC/UHPLC compatible.
  • USP classification: L7.