Learn More
Waters Corp Waters XBridge BEH C8 Column, 130Å, 2.5 μm, 2.1 mm x 50 mm
Supplier:  Waters Corp 176002555
Encompass_Preferred
The XBridge BEH C8 sorbent provides less retention than C18, enabling faster analysis with improved peak shape and chromatographic performance. This column features a trifunctionally bonded sorbent for a wide pH range (1 to 12), superior low pH stability, and ultra-low column bleed. Packed in ultra-low dispersion hardware, these 2.5 μm XP columns deliver exceptional separation, robustness, and throughput for HPLC assays, facilitating UPLC adoption.
  • Increased pH stability and longer column life.
  • Higher assay robustness due to improved column reliability.
  • High efficiency, symmetrical peak form, and exceptional pH stability across a broad pH range.
  • Unmatched repeatability, consistent performance from batch to batch and year to year.
Catalog No. 50-179-214