Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Waters XBridge BEH C8 XP Column, 130Å, 2.5 μm, 3 mm x 50 mm

Catalog No. 50179218 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

The XBridge BEH C8 sorbent is designed for method development, offering less retention than the XBridge BEH C18 sorbent, which can lead to faster separations. It improves peak shape and chromatographic performance and features a trifunctionally bonded sorbent with a wide usable pH range (1 to 12), superior low pH stability, and ultra-low column bleed. These 2.5 μm eXtended Performance (XP) columns, packed in ultra-low dispersion hardware, provide exceptional separation performance, robustness, and throughput for HPLC assays, while also facilitating a smooth transition to UPLC.

  • Chemistry: C8
  • Separation Mode: Reversed Phase
  • Particle Substrate: Hybrid
  • pH Range: 1 to 12
  • Maximum Pressure: 18000 psi (1240 Bar)
  • Particle Size: 2.5 μm
  • Pore Size: 130 Å
  • Inner Diameter: 3 mm
  • Length: 50 mm
  • Units per Package: 3 pk

Catalog No. 50-179-218 Supplier Waters Corp Supplier No. 176002559
May include imposed supplier surcharges.
Add to Cart
Add to Cart

The XBridge BEH C8 sorbent is designed for method development, offering less retention than the XBridge BEH C18 sorbent, which can lead to faster separations. It improves peak shape and chromatographic performance and features a trifunctionally bonded sorbent with a wide usable pH range (1 to 12), superior low pH stability, and ultra-low column bleed. These 2.5 μm eXtended Performance (XP) columns, packed in ultra-low dispersion hardware, provide exceptional separation performance, robustness, and throughput for HPLC assays, while also facilitating a smooth transition to UPLC.

  • Chemistry: C8
  • Separation Mode: Reversed Phase
  • Particle Substrate: Hybrid
  • pH Range: 1 to 12
  • Maximum Pressure: 18000 psi (1240 Bar)
  • Particle Size: 2.5 μm
  • Pore Size: 130 Å
  • Inner Diameter: 3 mm
  • Length: 50 mm
  • Units per Package: 3 pk

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.