Learn More
Waters Corp Waters VanGuard Cartridge, BEH C8, 130Å, 2.5 μm, 2.1 mm x 5 mm
Catalog No. 502038197 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This guard cartridge is designed to protect analytical columns and extend their lifespan by removing particulate contamination. It is optimized for 2.5 μm sorbent particles and can withstand high pressures, making it suitable for UPLC and UHPLC systems. It features a 130 Å pore size, hybrid particle substrate, and low silanol activity. This cartridge has an inner diameter of 2.1 mm and a length of 5 mm.
  • Extends analytical column lifetime and performance.
  • Removes particulate contamination from mobile phase.
  • Designed for 2.5 μm sorbent particles.
  • Operates within a pH range of 1 to 12.
  • Withstands up to 18,000 psi (1240 Bar) pressure.
  • Features 130 Å pore size.
  • Inner diameter of 2.1 mm and length of 5 mm.
  • Designed for UPLC and UHPLC systems.
Catalog No. 50-203-8197 Supplier Waters Corp Supplier No. 186007781
This guard cartridge is designed to protect analytical columns and extend their lifespan by removing particulate contamination. It is optimized for 2.5 μm sorbent particles and can withstand high pressures, making it suitable for UPLC and UHPLC systems. It features a 130 Å pore size, hybrid particle substrate, and low silanol activity. This cartridge has an inner diameter of 2.1 mm and a length of 5 mm.
  • Extends analytical column lifetime and performance.
  • Removes particulate contamination from mobile phase.
  • Designed for 2.5 μm sorbent particles.
  • Operates within a pH range of 1 to 12.
  • Withstands up to 18,000 psi (1240 Bar) pressure.
  • Features 130 Å pore size.
  • Inner diameter of 2.1 mm and length of 5 mm.
  • Designed for UPLC and UHPLC systems.