Learn More
Waters Corp XBridge BEH HILIC Method Validation Kit, 130Å, 2.5 µm, 3 mm X 150 mm, 3/pk
Catalog No. 50466472 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This method validation kit consists of three XBridge BEH HILIC columns packed with three different batches of sorbent. XBridge BEH HILIC columns contain unbonded BEH substrate and are designed to retain and separate very polar compounds. These unique columns are optimized and tested to produce efficient, reproducible separations under HPLC HILIC conditions. More rugged than silica based HILIC phases, the BEH particle’s chemical stability results in long column lifetimes. Packed in ultra-low dispersion hardware, 2.5 µm eXtended Performance [XP] columns enable exceptional separation performance, robustness and throughput for HPLC assays while enabling a seamless transition path toward UPLC adoption.
Catalog No. 50-466-472 Supplier Waters Corp Supplier No. 186006780
This method validation kit consists of three XBridge BEH HILIC columns packed with three different batches of sorbent. XBridge BEH HILIC columns contain unbonded BEH substrate and are designed to retain and separate very polar compounds. These unique columns are optimized and tested to produce efficient, reproducible separations under HPLC HILIC conditions. More rugged than silica based HILIC phases, the BEH particle’s chemical stability results in long column lifetimes. Packed in ultra-low dispersion hardware, 2.5 µm eXtended Performance [XP] columns enable exceptional separation performance, robustness and throughput for HPLC assays while enabling a seamless transition path toward UPLC adoption.