Learn More
Waters Corp Waters XBridge BEH HILIC Column, 130Å, 2.5 μm, 3 mm X 150 mm
Catalog No. 50466430 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
XBridge BEH HILIC columns utilize unbonded BEH substrate for retaining and separating very polar compounds. Optimized for efficient, reproducible HPLC HILIC separations, these columns offer ruggedness and extended lifetimes due to BEH particle chemical stability. Featuring ultra-low dispersion hardware, 2.5 μm XP columns deliver exceptional performance and throughput for HPLC assays, supporting seamless UPLC adoption.
  • Retains and separates very polar compounds.
  • Ensures efficient, reproducible HPLC HILIC separations.
  • Provides ruggedness and long column lifetimes.
  • Features ultra-low dispersion hardware.
  • Delivers exceptional performance and throughput for HPLC assays.
  • Supports seamless UPLC adoption.
Catalog No. 50-466-430 Supplier Waters Corp Supplier No. 186006722
XBridge BEH HILIC columns utilize unbonded BEH substrate for retaining and separating very polar compounds. Optimized for efficient, reproducible HPLC HILIC separations, these columns offer ruggedness and extended lifetimes due to BEH particle chemical stability. Featuring ultra-low dispersion hardware, 2.5 μm XP columns deliver exceptional performance and throughput for HPLC assays, supporting seamless UPLC adoption.
  • Retains and separates very polar compounds.
  • Ensures efficient, reproducible HPLC HILIC separations.
  • Provides ruggedness and long column lifetimes.
  • Features ultra-low dispersion hardware.
  • Delivers exceptional performance and throughput for HPLC assays.
  • Supports seamless UPLC adoption.