Learn More
Waters Corp Waters XBridge BEH HILIC Column, 2.5 μm, 4.6 x 150 mm
Catalog No. 50466431 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
XBridge BEH HILIC columns are part of the eXtended Performance (XP) 2.5 μm series. Designed for seamless transition between HPLC and UPLC™ Technology, they are compatible with any LC system, offering flexibility and improved productivity at intermediate backpressures.
  • Enhances transition strategy between HPLC and UPLC™ technology.
  • Compatible with all LC systems (HPLC, UHPLC, UPLC) for improved productivity.
  • Offers diverse selectivity with multiple particle substrates and chemistries.
  • Provides 40% lower backpressure than 1.7 μm columns, ensuring superior performance and throughput.
  • Enables rapid method optimization and future-proof method transfer.
  • Utilizes advanced packing technology for exceptional performance, loading capacity, and for complex mixture analysis.
Catalog No. 50-466-431 Supplier Waters Corp Supplier No. 186006723
XBridge BEH HILIC columns are part of the eXtended Performance (XP) 2.5 μm series. Designed for seamless transition between HPLC and UPLC™ Technology, they are compatible with any LC system, offering flexibility and improved productivity at intermediate backpressures.
  • Enhances transition strategy between HPLC and UPLC™ technology.
  • Compatible with all LC systems (HPLC, UHPLC, UPLC) for improved productivity.
  • Offers diverse selectivity with multiple particle substrates and chemistries.
  • Provides 40% lower backpressure than 1.7 μm columns, ensuring superior performance and throughput.
  • Enables rapid method optimization and future-proof method transfer.
  • Utilizes advanced packing technology for exceptional performance, loading capacity, and for complex mixture analysis.