Learn More
Waters Corp HPLC Column, BEH HILIC, 10 x 100 mm, 5 μm, 130 Å
Catalog No. 502391594 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
XBridge BEH HILIC columns, with an unbound BEH substrate, retain and separate highly polar compounds. Optimized for efficient HPLC HILIC separations, their robust BEH particles and chemical stability ensure a long column lifetime. This column has a 10 mm inner diameter, 100 mm length, and 5 μm particle size.
  • Retains and separates highly polar compounds.
  • Optimized for efficient HPLC HILIC separations.
  • Robust BEH particles provide increased stability and long column lifetime.
  • BEH (Ethylene Bridge Hybrid Particle) technology.
  • Withstands high pH, high temperatures, and extreme operating conditions.
  • Suitable for LC-MS analyses of small molecules and biomolecules.
  • pH range of 1 to 9.
  • Medium silanol activity.
  • Surface area of 185 m²/g.
  • Spherical particles.
  • Pore size of 130 Å.
Catalog No. 50-239-1594 Supplier Waters Corp Supplier No. 186008218
XBridge BEH HILIC columns, with an unbound BEH substrate, retain and separate highly polar compounds. Optimized for efficient HPLC HILIC separations, their robust BEH particles and chemical stability ensure a long column lifetime. This column has a 10 mm inner diameter, 100 mm length, and 5 μm particle size.
  • Retains and separates highly polar compounds.
  • Optimized for efficient HPLC HILIC separations.
  • Robust BEH particles provide increased stability and long column lifetime.
  • BEH (Ethylene Bridge Hybrid Particle) technology.
  • Withstands high pH, high temperatures, and extreme operating conditions.
  • Suitable for LC-MS analyses of small molecules and biomolecules.
  • pH range of 1 to 9.
  • Medium silanol activity.
  • Surface area of 185 m²/g.
  • Spherical particles.
  • Pore size of 130 Å.