Learn More
Waters Corp Waters Prep Guard Cartridge, 130Å, 5 μm, 30 mm x 10 mm
Catalog No. NC1137390 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The 30x10mm Prep Guard Cartridge, used with the 30x10mm Prep Guard Holder, is designed to safeguard both HPLC and SFC 30 mm ID Prep OBD™ columns. Its placement ahead of the prep column helps extend column lifetimes and reduce purification costs.
  • Protects HPLC and SFC 30 mm ID prep OBD™ columns.
  • Aids in achieving longer column lifetimes.
  • Contributes to reduced purification costs.
Catalog No. NC1137390 Supplier Waters Corp Supplier No. 186006896
The 30x10mm Prep Guard Cartridge, used with the 30x10mm Prep Guard Holder, is designed to safeguard both HPLC and SFC 30 mm ID Prep OBD™ columns. Its placement ahead of the prep column helps extend column lifetimes and reduce purification costs.
  • Protects HPLC and SFC 30 mm ID prep OBD™ columns.
  • Aids in achieving longer column lifetimes.
  • Contributes to reduced purification costs.