Learn More
Waters Corp XBridge BEH HILIC VanGuard Cartridge, 130Å, 3.5 μm, 2.1 mm × 5 mm
Catalog No. 502390552 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
XBridge BEH HILIC VanGuard Cartridges are designed to extend the lifetime and performance of analytical columns by removing particulate contamination. They protect 2.1 mm and 3.0 mm I.D. HILIC analytical columns with 3.5 μm sorbent particles, improving analytical performance and preserving separation efficiency under high pressures.
  • Extends analytical column lifetime and performance by removing particulate contamination.
  • Optimized to protect 2.1 mm and 3.0 mm I.D. analytical columns with 3.5 μm sorbent particles.
  • Easy to install and practical to use.
  • Preserves UPLC, UHPLC, and HPLC separation efficiency.
  • Withstands high pressures in ACQUITY UPLC System applications.
  • Offers a broad pH range of 1–9.
Catalog No. 50-239-0552 Supplier Waters Corp Supplier No. 186007784
XBridge BEH HILIC VanGuard Cartridges are designed to extend the lifetime and performance of analytical columns by removing particulate contamination. They protect 2.1 mm and 3.0 mm I.D. HILIC analytical columns with 3.5 μm sorbent particles, improving analytical performance and preserving separation efficiency under high pressures.
  • Extends analytical column lifetime and performance by removing particulate contamination.
  • Optimized to protect 2.1 mm and 3.0 mm I.D. analytical columns with 3.5 μm sorbent particles.
  • Easy to install and practical to use.
  • Preserves UPLC, UHPLC, and HPLC separation efficiency.
  • Withstands high pressures in ACQUITY UPLC System applications.
  • Offers a broad pH range of 1–9.