Learn More
Waters Corp VanGuard Cartridge, HILIC, 130 Å, 5 μm
Catalog No. 502391067 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These VanGuard Cartridges are designed to extend the lifetime and performance of analytical columns by removing particulate and chemical contamination from the mobile phase stream. This cartridge is optimized for protecting 4.6 mm I.D. XBridge BEH HILIC analytical columns that contain 5 μm sorbent particles.
  • Prolongs the lifetime of analytical columns
  • Removes particulates and chemical contamination
  • Maintains UPLC™, UHPLC, and HPLC separation efficiency
  • Provides cost-effective protection for analytical columns
Catalog No. 50-239-1067 Supplier Waters Corp Supplier No. 186007789
These VanGuard Cartridges are designed to extend the lifetime and performance of analytical columns by removing particulate and chemical contamination from the mobile phase stream. This cartridge is optimized for protecting 4.6 mm I.D. XBridge BEH HILIC analytical columns that contain 5 μm sorbent particles.
  • Prolongs the lifetime of analytical columns
  • Removes particulates and chemical contamination
  • Maintains UPLC™, UHPLC, and HPLC separation efficiency
  • Provides cost-effective protection for analytical columns