Learn More
Waters Corp Waters Column, BEH HILIC, 2.5 μm, 3 mm X 50 mm
Supplier:  Waters Corp 176002583
Encompass_Preferred
These BEH HILIC columns, with unbonded BEH substrate, are designed for the retention and separation of very polar compounds. Optimized and tested for efficient, reproducible separations under HPLC HILIC conditions, they offer greater ruggedness than silica-based HILIC phases due to the chemical stability of the BEH particle, ensuring long column lifetimes. Packed in ultra-low dispersion hardware, the 2.5 μm eXtended Performance columns provide exceptional separation performance, robustness, and throughput for HPLC assays, while facilitating seamless UPLC adoption.
  • More rugged than silica-based HILIC phases due to BEH particle chemical stability, extending column lifetimes.
  • eXtended performance columns with 2.5 μm particles deliver exceptional separation performance and throughput for HPLC assays.
  • Provides a seamless transition path toward UPLC adoption.
  • Contains unbonded BEH substrate.
Catalog No. 50-179-242