Learn More
Waters Corp Waters XBridge BEH HILIC XP Column, 2.5 μm, 2.1 x 30 mm
Supplier:  Waters Corp 186006076
Encompass_Preferred
eXtended Performance [XP] 2.5 μm Columns are designed to enhance the transition strategy between HPLC and UPLC™ Technology. These columns are compatible with any LC system and offer flexibility to HPLC, UHPLC, and UPLC users to improve productivity at intermediate backpressures. They feature a diverse range of stationary phases that are fully and seamlessly scalable between HPLC and UPLC platforms.
  • Compatible with any LC system
  • Offers flexibility to HPLC, UHPLC, and UPLC users
  • Improves productivity at intermediate backpressures
  • Features a diverse range of stationary phases
  • Fully and seamlessly scalable between HPLC and UPLC platforms
Catalog No. 50-179-498