Learn More
Waters Corp Waters XBridge BEH HILIC Column, 130Å, 2.5 μm, 3 mm × 50 mm
Catalog No. 50179503 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
XBridge BEH HILIC columns contain unbonded BEH substrate, designed for efficient, reproducible separations of very polar compounds under HPLC HILIC conditions. More rugged than silica-based phases, the BEH particle’s chemical stability ensures long column lifetimes. These 2.5 μm eXtended Performance [XP] columns provide exceptional separation performance, robustness, and throughput for HPLC assays, facilitating a seamless transition to UPLC adoption.
  • HILIC chemistry
  • Hybrid particle substrate
  • pH range of 1 to 9
  • Maximum pressure of 18000 psi (1240 Bar)
  • No endcapping
  • Low silanol activity
  • Spherical particle shape
  • 2.5 μm particle size
  • Parker-style endfitting type
  • 130 Å pore size
  • Compatible with UPLC and UHPLC systems
  • USP classification L3
  • 3 mm inner diameter
  • 50 mm length
  • 1 unit per package
Catalog No. 50-179-503 Supplier Waters Corp Supplier No. 186006081
XBridge BEH HILIC columns contain unbonded BEH substrate, designed for efficient, reproducible separations of very polar compounds under HPLC HILIC conditions. More rugged than silica-based phases, the BEH particle’s chemical stability ensures long column lifetimes. These 2.5 μm eXtended Performance [XP] columns provide exceptional separation performance, robustness, and throughput for HPLC assays, facilitating a seamless transition to UPLC adoption.
  • HILIC chemistry
  • Hybrid particle substrate
  • pH range of 1 to 9
  • Maximum pressure of 18000 psi (1240 Bar)
  • No endcapping
  • Low silanol activity
  • Spherical particle shape
  • 2.5 μm particle size
  • Parker-style endfitting type
  • 130 Å pore size
  • Compatible with UPLC and UHPLC systems
  • USP classification L3
  • 3 mm inner diameter
  • 50 mm length
  • 1 unit per package