Learn More
Waters Corp Waters XBridge BEH HILIC XP Column, 130Å, 2.5 μm, 3 mm × 100 mm
Catalog No. 50179244 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
XBridge BEH HILIC XP Columns are designed for the retention and separation of very polar compounds, offering efficient and reproducible results for HPLC assays.
  • Contain unbonded BEH substrate for separating very polar compounds.
  • Optimized and tested for efficient, reproducible separations under HPLC HILIC conditions.
  • More rugged than silica-based HILIC phases due to BEH particle's chemical stability.
  • Ensures long column lifetimes.
  • Packed in ultra-low dispersion hardware.
  • 2.5 μm eXtended Performance [XP] columns.
  • Enables exceptional separation performance, robustness, and throughput for HPLC assays.
  • Provides a seamless transition path toward UPLC adoption.
  • Maximum pressure of 18000 psi (1240 Bar).
  • Pore size is 130 Å.
Catalog No. 50-179-244 Supplier Waters Corp Supplier No. 176002585
XBridge BEH HILIC XP Columns are designed for the retention and separation of very polar compounds, offering efficient and reproducible results for HPLC assays.
  • Contain unbonded BEH substrate for separating very polar compounds.
  • Optimized and tested for efficient, reproducible separations under HPLC HILIC conditions.
  • More rugged than silica-based HILIC phases due to BEH particle's chemical stability.
  • Ensures long column lifetimes.
  • Packed in ultra-low dispersion hardware.
  • 2.5 μm eXtended Performance [XP] columns.
  • Enables exceptional separation performance, robustness, and throughput for HPLC assays.
  • Provides a seamless transition path toward UPLC adoption.
  • Maximum pressure of 18000 psi (1240 Bar).
  • Pore size is 130 Å.