Learn More
Waters Corp Waters Corporation Column, HILIC, 2.5 μm, 2.1 mm X 50 mm, 130Å
Catalog No. 50179499 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These HILIC columns efficiently separate very polar compounds under HPLC conditions. Offering superior ruggedness and extended lifetimes due to chemical stability, the 2.5 μm eXtended Performance [XP] columns are packed in ultra-low dispersion hardware. This ensures exceptional performance, robustness, and throughput, facilitating a seamless transition to UPLC.
  • Retains and separates very polar compounds using unbonded hybrid substrate.
  • Optimized for efficient, reproducible separations under HPLC HILIC.
  • More rugged with longer lifetimes due to chemical stability.
  • 2.5 μm XP columns provide exceptional performance and robustness.
  • Enables seamless UPLC adoption.
  • Enhances retention for polar analytes via partitioning, ion exchange, and hydrogen bonding.
  • Offers complementary and alternate selectivity to reversed-phase for orthogonal separations.
Catalog No. 50-179-499 Supplier Waters Corp Supplier No. 186006077
These HILIC columns efficiently separate very polar compounds under HPLC conditions. Offering superior ruggedness and extended lifetimes due to chemical stability, the 2.5 μm eXtended Performance [XP] columns are packed in ultra-low dispersion hardware. This ensures exceptional performance, robustness, and throughput, facilitating a seamless transition to UPLC.
  • Retains and separates very polar compounds using unbonded hybrid substrate.
  • Optimized for efficient, reproducible separations under HPLC HILIC.
  • More rugged with longer lifetimes due to chemical stability.
  • 2.5 μm XP columns provide exceptional performance and robustness.
  • Enables seamless UPLC adoption.
  • Enhances retention for polar analytes via partitioning, ion exchange, and hydrogen bonding.
  • Offers complementary and alternate selectivity to reversed-phase for orthogonal separations.