Learn More
Waters Corp Waters Column, 130Å, 2.5 μm, 2.1 mm × 100 mm
Catalog No. 50179240 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
XBridge BEH HILIC columns contain unbonded BEH substrate and are designed to retain and separate very polar compounds. More rugged than silica-based HILIC phases, the BEH particle's chemical stability results in long column lifetimes. Packed in ultra-low dispersion hardware, 2.5 μm eXtended Performance [XP] columns enable exceptional separation performance, robustness, and throughput for HPLC assays.
  • Designed to retain and separate very polar compounds.
  • Optimized and tested to produce efficient, reproducible separations under HPLC HILIC conditions.
  • More rugged than silica-based HILIC phases, with the BEH particle's chemical stability leading to long column lifetimes.
  • Packed in ultra-low dispersion hardware.
  • Provides exceptional separation performance, robustness, and throughput for HPLC assays.
  • Enables a seamless transition path toward UPLC adoption.
Catalog No. 50-179-240 Supplier Waters Corp Supplier No. 176002581
XBridge BEH HILIC columns contain unbonded BEH substrate and are designed to retain and separate very polar compounds. More rugged than silica-based HILIC phases, the BEH particle's chemical stability results in long column lifetimes. Packed in ultra-low dispersion hardware, 2.5 μm eXtended Performance [XP] columns enable exceptional separation performance, robustness, and throughput for HPLC assays.
  • Designed to retain and separate very polar compounds.
  • Optimized and tested to produce efficient, reproducible separations under HPLC HILIC conditions.
  • More rugged than silica-based HILIC phases, with the BEH particle's chemical stability leading to long column lifetimes.
  • Packed in ultra-low dispersion hardware.
  • Provides exceptional separation performance, robustness, and throughput for HPLC assays.
  • Enables a seamless transition path toward UPLC adoption.