Learn More
Waters Corp XBridge BEH HILIC XP Column, 130Å, 2.5 µm, 2.1 mm X 30 mm, 3/pk
Catalog No. 50179237 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
XBridge BEH HILIC columns contain unbonded BEH substrate and are designed to retain and separate very polar compounds. These unique columns are optimized and tested to produce efficient, reproducible separations under HPLC HILIC conditions. More rugged than silica based HILIC phases, the BEH particle’s chemical stability results in long column lifetimes. Packed in ultra-low dispersion hardware, 2.5 µm eXtended Performance [XP] columns enable exceptional separation performance, robustness and throughput for HPLC assays while enabling a seamless transition path toward UPLC adoption.
Catalog No. 50-179-237 Supplier Waters Corp Supplier No. 176002578
XBridge BEH HILIC columns contain unbonded BEH substrate and are designed to retain and separate very polar compounds. These unique columns are optimized and tested to produce efficient, reproducible separations under HPLC HILIC conditions. More rugged than silica based HILIC phases, the BEH particle’s chemical stability results in long column lifetimes. Packed in ultra-low dispersion hardware, 2.5 µm eXtended Performance [XP] columns enable exceptional separation performance, robustness and throughput for HPLC assays while enabling a seamless transition path toward UPLC adoption.