Learn More
Waters Corp Waters XBridge BEH HILIC Column, 130Å, 2.5 μm, 3 mm X 30 mm
Catalog No. 50179502 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Waters XBridge BEH HILIC Column, 130Å, 2.5 μm, 3 mm X 30 mm
Catalog No. 50-179-502 Supplier Waters Corp Supplier No. 186006080
Waters XBridge BEH HILIC Column, 130Å, 2.5 μm, 3 mm X 30 mm