Learn More
Waters Corp Waters Corporation Column, XBridge BEH HILIC, 130Å, 2.5 μm, 3 mm X 75 mm
Supplier:  Waters Corp 186006082
Encompass_Preferred
XBridge BEH HILIC columns effectively retain and separate very polar compounds. Optimized for reproducible HPLC HILIC separations, these columns utilize unbonded BEH substrate for enhanced ruggedness and chemical stability, ensuring long column lifetimes. With ultra-low dispersion hardware, 2.5 μm XP columns offer exceptional performance and throughput for HPLC assays, facilitating a seamless transition to UPLC.
  • Unbonded BEH substrate for very polar compound retention and separation.
  • Optimized for efficient, reproducible HPLC HILIC separations.
  • Rugged design with BEH particle chemical stability for extended column lifetimes.
  • Ultra-low dispersion hardware and 2.5 μm XP particles ensure exceptional performance and high throughput.
  • Facilitates seamless transition to UPLC adoption.
Catalog No. 50-179-504