Learn More
Waters Corp Waters Column, BEH HILIC, 2.5 μm, 4.6 mm × 30 mm, 130 Å
Catalog No. 50201530 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These BEH HILIC columns utilize an unbonded BEH substrate, designed for retaining and separating highly polar compounds. They provide efficient, reproducible HPLC HILIC separations with greater ruggedness and extended lifetimes due to BEH particle chemical stability. The 2.5 μm eXtended Performance (XP) columns, packed in ultra-low dispersion hardware, offer exceptional separation performance and throughput for HPLC, enabling smooth UPLC transition.
  • Retains and separates very polar compounds.
  • Optimized for efficient, reproducible HPLC HILIC separations.
  • Rugged and long-lasting due to BEH particle chemical stability.
  • Packed in ultra-low dispersion hardware for 2.5 μm XP.
  • Exceptional separation performance, robustness, and throughput for HPLC.
  • Seamless transition path toward UPLC adoption.
Catalog No. 50-201-530 Supplier Waters Corp Supplier No. 186006084
These BEH HILIC columns utilize an unbonded BEH substrate, designed for retaining and separating highly polar compounds. They provide efficient, reproducible HPLC HILIC separations with greater ruggedness and extended lifetimes due to BEH particle chemical stability. The 2.5 μm eXtended Performance (XP) columns, packed in ultra-low dispersion hardware, offer exceptional separation performance and throughput for HPLC, enabling smooth UPLC transition.
  • Retains and separates very polar compounds.
  • Optimized for efficient, reproducible HPLC HILIC separations.
  • Rugged and long-lasting due to BEH particle chemical stability.
  • Packed in ultra-low dispersion hardware for 2.5 μm XP.
  • Exceptional separation performance, robustness, and throughput for HPLC.
  • Seamless transition path toward UPLC adoption.