Learn More
Waters Corp XBridge BEH HILIC Method Validation Kit, 130Å, 2.5 µm, 3 mm X 100 mm, 3/pk
Catalog No. 50179595 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This method validation kit consists of three batches of XBridge BEH HILIC columns packed with three different batches of sorbent
Catalog No. 50-179-595 Supplier Waters Corp Supplier No. 186006224
Add to Cart
Add to Cart
This method validation kit consists of three batches of XBridge BEH HILIC columns packed with three different batches of sorbent