Learn More
Waters Corp XBridge BEH HILIC XP Column, 4.6 mm x 30 mm, 2.5 μm
Catalog No. 50201531 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
XBridge BEH HILIC XP columns are designed for highly polar compounds. They use an unbonded BEH substrate for chemical stability and ruggedness, optimizing HPLC HILIC separations and facilitating UPLC adoption.
  • Designed for retention and separation of very polar compounds.
  • Contains an unbonded BEH (Ethylene Bridged Hybrid) substrate, offering chemical stability and ruggedness.
  • Optimized for efficient, reproducible separations under HPLC HILIC conditions.
  • eXtended Performance (XP) columns with 2.5 μm particles deliver exceptional separation performance.
  • Enables a seamless transition path toward UPLC adoption.
  • Operates within a pH range of 1 to 9.
Catalog No. 50-201-531 Supplier Waters Corp Supplier No. 186006085
XBridge BEH HILIC XP columns are designed for highly polar compounds. They use an unbonded BEH substrate for chemical stability and ruggedness, optimizing HPLC HILIC separations and facilitating UPLC adoption.
  • Designed for retention and separation of very polar compounds.
  • Contains an unbonded BEH (Ethylene Bridged Hybrid) substrate, offering chemical stability and ruggedness.
  • Optimized for efficient, reproducible separations under HPLC HILIC conditions.
  • eXtended Performance (XP) columns with 2.5 μm particles deliver exceptional separation performance.
  • Enables a seamless transition path toward UPLC adoption.
  • Operates within a pH range of 1 to 9.