Learn More
Waters Corp Waters XBridge BEH HILIC XP Column, 4.6 X 100 mm, 2.5 μm
Catalog No. 50201533 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
XBridge BEH HILIC columns utilize an unbonded BEH substrate for retaining and separating very polar compounds. Optimized for efficient, reproducible HPLC HILIC separations, they offer superior ruggedness and long column lifetimes due to BEH particle chemical stability. The 2.5 μm XP columns are packed in ultra-low dispersion hardware, ensuring exceptional separation performance, robustness, and throughput for HPLC assays, and facilitate a smooth transition to UPLC.
  • Retains and separates very polar compounds.
  • Provides efficient, reproducible HPLC HILIC separations.
  • Ensures long column lifetimes and ruggedness with BEH particle stability.
Catalog No. 50-201-533 Supplier Waters Corp Supplier No. 186006087
Add to Cart
Add to Cart
XBridge BEH HILIC columns utilize an unbonded BEH substrate for retaining and separating very polar compounds. Optimized for efficient, reproducible HPLC HILIC separations, they offer superior ruggedness and long column lifetimes due to BEH particle chemical stability. The 2.5 μm XP columns are packed in ultra-low dispersion hardware, ensuring exceptional separation performance, robustness, and throughput for HPLC assays, and facilitate a smooth transition to UPLC.
  • Retains and separates very polar compounds.
  • Provides efficient, reproducible HPLC HILIC separations.
  • Ensures long column lifetimes and ruggedness with BEH particle stability.