Learn More
Waters Corp XBridge BEH HILIC XP VanGuard Cartridge, 130Å, 2.5 μm, 2.1 mm x 5 mm
Catalog No. 502390764 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
XBridge BEH HILIC XP VanGuard Cartridges extend analytical column lifetime and performance by removing particulate and chemical contamination from the mobile phase. Optimized to protect 2.1 mm and 3.0 mm I.D. XBridge BEH HILIC analytical columns with 2.5 μm sorbent particles, this cartridge is easy to use, install, and capable of sustaining high pressures in applications run on the ACQUITY UPLC System.
  • Extends analytical column lifetime and performance.
  • Removes particulate and chemical contamination from the mobile phase stream.
  • Optimized to protect 2.1 mm and 3.0 mm I.D. XBridge BEH HILIC analytical columns with 2.5 μm sorbent particles.
  • Easy to use and install.
  • Can sustain high pressures in applications run on the ACQUITY UPLC System.
  • Features BEH Technology C18, 130-pore, and 2.5 μm particles.
  • Quality controlled and tested using a peptide map.
Catalog No. 50-239-0764 Supplier Waters Corp Supplier No. 186007790
XBridge BEH HILIC XP VanGuard Cartridges extend analytical column lifetime and performance by removing particulate and chemical contamination from the mobile phase. Optimized to protect 2.1 mm and 3.0 mm I.D. XBridge BEH HILIC analytical columns with 2.5 μm sorbent particles, this cartridge is easy to use, install, and capable of sustaining high pressures in applications run on the ACQUITY UPLC System.
  • Extends analytical column lifetime and performance.
  • Removes particulate and chemical contamination from the mobile phase stream.
  • Optimized to protect 2.1 mm and 3.0 mm I.D. XBridge BEH HILIC analytical columns with 2.5 μm sorbent particles.
  • Easy to use and install.
  • Can sustain high pressures in applications run on the ACQUITY UPLC System.
  • Features BEH Technology C18, 130-pore, and 2.5 μm particles.
  • Quality controlled and tested using a peptide map.