Learn More
Waters Corp XBridge BEH Phenyl XP Column, 130Å, 2.5 μm, 2.1 mm × 150 mm
Supplier:  Waters Corp 176002885
Encompass_Preferred
XBridge BEH Phenyl columns offer complementary selectivity to C18 stationary phases, especially for polyaromatic compounds. With the most chemically stable Phenyl sorbent, they ensure ultra-low column bleed, long lifetimes, and excellent peak shape. Packed in ultra-low dispersion hardware, 2.5 μm XP columns deliver exceptional separation and throughput for HPLC assays, enabling a seamless transition to UPLC.
  • Offers complementary selectivity for polyaromatic compounds.
  • Most chemically stable Phenyl sorbent; ultra-low bleed and long lifetimes.
  • Exceptional separation, robustness, and throughput with 2.5 μm XP columns.
  • Seamless HPLC to UPLC assay transition.
  • Wider pH range (1 to 12 pH) with outstanding stability.
  • Tolerates up to 10,000 psi (690 bar) operating pressure.
Catalog No. 50-465-925