Learn More
Waters Corp Waters XBridge BEH Phenyl XP Column, 130Å, 2.5 μm, 2.1 mm x 150 mm
Catalog No. 50466426 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
XBridge BEH Phenyl columns offer selectivity complementary to C18 stationary phases, particularly for polyaromatic compounds. This phenyl sorbent ensures ultra-low column bleed, long lifetimes, and excellent peak shape, enabling high performance HPLC assays and seamless UPLC adoption.
  • Complementary selectivity to C18 stationary phases, ideal for polyaromatic compounds.
  • Chemically stable phenyl sorbent with ultra-low column bleed and long column lifetimes.
  • 2.5 μm XP columns provide exceptional separation performance and robustness.
  • Enables a seamless transition path toward UPLC adoption.
  • Unmatched column longevity, functioning across a wide pH (1 to 12 pH) and temperature range.
  • Broader pH range compared to standard phenyl liquid chromatography columns for robust method development.
Catalog No. 50-466-426 Supplier Waters Corp Supplier No. 186006718
XBridge BEH Phenyl columns offer selectivity complementary to C18 stationary phases, particularly for polyaromatic compounds. This phenyl sorbent ensures ultra-low column bleed, long lifetimes, and excellent peak shape, enabling high performance HPLC assays and seamless UPLC adoption.
  • Complementary selectivity to C18 stationary phases, ideal for polyaromatic compounds.
  • Chemically stable phenyl sorbent with ultra-low column bleed and long column lifetimes.
  • 2.5 μm XP columns provide exceptional separation performance and robustness.
  • Enables a seamless transition path toward UPLC adoption.
  • Unmatched column longevity, functioning across a wide pH (1 to 12 pH) and temperature range.
  • Broader pH range compared to standard phenyl liquid chromatography columns for robust method development.