Learn More
Waters Corp Waters XBridge BEH Phenyl Method Validation Kit, 2.5 μm, 2.1 mm × 150 mm, 3 pk
Catalog No. 50466468 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This method validation kit includes three XBridge BEH Phenyl columns with different sorbent batches. They offer complementary selectivity to C18 phases, superior stability, and excellent performance for HPLC and UPLC applications. Documentation available online.
  • Includes three columns for method validation.
  • Phenyl chemistry with complementary C18 selectivity.
  • Superior chemical stability, ultra-low column bleed, and long lifetimes.
  • 2.5 μm BEH particle technology for exceptional HPLC/UPLC performance.
  • pH range: 1 to 12; Max pressure: 18000 psi (1240 Bar).
  • Pore size: 130 Å; Dimensions: 2.1 mm ID × 150 mm L.
Catalog No. 50-466-468 Supplier Waters Corp Supplier No. 186006776
This method validation kit includes three XBridge BEH Phenyl columns with different sorbent batches. They offer complementary selectivity to C18 phases, superior stability, and excellent performance for HPLC and UPLC applications. Documentation available online.
  • Includes three columns for method validation.
  • Phenyl chemistry with complementary C18 selectivity.
  • Superior chemical stability, ultra-low column bleed, and long lifetimes.
  • 2.5 μm BEH particle technology for exceptional HPLC/UPLC performance.
  • pH range: 1 to 12; Max pressure: 18000 psi (1240 Bar).
  • Pore size: 130 Å; Dimensions: 2.1 mm ID × 150 mm L.