Learn More
Waters Corp Method Validation Kit, Phenyl, 130Å, 2.5μm, 3.0 mm ID, 150 mm Length
Catalog No. 50466469 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This Method Validation Kit features a phenyl phase and is designed with a 130Å pore size and 2.5μm particle size. It has dimensions of 3.0 mm ID and 150 mm length, suitable for various analytical applications.
  • Identified as a Method Validation Kit
  • 3.0 mm ID
  • 150 mm length
  • 2.5 μm particle size
  • Phenyl phase type
  • 130 Å pore size
  • USP L11 compliant
Catalog No. 50-466-469 Supplier Waters Corp Supplier No. 186006777
This Method Validation Kit features a phenyl phase and is designed with a 130Å pore size and 2.5μm particle size. It has dimensions of 3.0 mm ID and 150 mm length, suitable for various analytical applications.
  • Identified as a Method Validation Kit
  • 3.0 mm ID
  • 150 mm length
  • 2.5 μm particle size
  • Phenyl phase type
  • 130 Å pore size
  • USP L11 compliant