Learn More
Waters Corp XBridge BEH Phenyl Method Validation Kit, 130Å, 2.5 µm, 4.6 mm X 150 mm, 3/pk
Catalog No. 50466470 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This method validation kit consists of three XBridge BEH Phenyl columns packed with three different batches of sorbent. XBridge BEH Phenyl columns provide complementary selectivity to C18 stationary phases, especially for polyaromatic compounds. This most chemically stable commercially available Phenyl sorbent on the market and provides you with ultra-low column bleed, long column lifetimes and excellent peak shape. Packed in ultra-low dispersion hardware, 2.5 µm eXtended Performance [XP] columns enable exceptional separation performance, robustness and throughput for HPLC assays while enabling a seamless transition path toward UPLC adoption.
Catalog No. 50-466-470 Supplier Waters Corp Supplier No. 186006778
This method validation kit consists of three XBridge BEH Phenyl columns packed with three different batches of sorbent. XBridge BEH Phenyl columns provide complementary selectivity to C18 stationary phases, especially for polyaromatic compounds. This most chemically stable commercially available Phenyl sorbent on the market and provides you with ultra-low column bleed, long column lifetimes and excellent peak shape. Packed in ultra-low dispersion hardware, 2.5 µm eXtended Performance [XP] columns enable exceptional separation performance, robustness and throughput for HPLC assays while enabling a seamless transition path toward UPLC adoption.