Learn More
Waters Corp Waters Column, Phenyl, 2.5 µm, 3 mm, 150 mm
Catalog No. 50466427 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These BEH Phenyl columns offer complementary selectivity to C18 stationary phases, particularly for polyaromatic compounds. They feature a chemically stable Phenyl sorbent, providing ultra-low column bleed, extended column lifetimes, and excellent peak shape. Ultra-low dispersion hardware (2.5 µm eXtended Performance [XP] columns) ensures exceptional separation performance for HPLC assays and facilitates a seamless transition to UPLC. This column also offers alternative selectivity for aromatic molecules and operates across a wide pH range of 1 to 12 and a broad temperature range.
  • Complementary selectivity for polyaromatic compounds with extended column lifetimes.
  • Ultra-low dispersion hardware for exceptional separation performance in HPLC.
  • Wide pH range (1 to 12) for robust method development.
  • Phenyl hexyl-trifunctional ligand ensures high pH stability and alternate selectivity.
Catalog No. 50-466-427 Supplier Waters Corp Supplier No. 186006719
These BEH Phenyl columns offer complementary selectivity to C18 stationary phases, particularly for polyaromatic compounds. They feature a chemically stable Phenyl sorbent, providing ultra-low column bleed, extended column lifetimes, and excellent peak shape. Ultra-low dispersion hardware (2.5 µm eXtended Performance [XP] columns) ensures exceptional separation performance for HPLC assays and facilitates a seamless transition to UPLC. This column also offers alternative selectivity for aromatic molecules and operates across a wide pH range of 1 to 12 and a broad temperature range.
  • Complementary selectivity for polyaromatic compounds with extended column lifetimes.
  • Ultra-low dispersion hardware for exceptional separation performance in HPLC.
  • Wide pH range (1 to 12) for robust method development.
  • Phenyl hexyl-trifunctional ligand ensures high pH stability and alternate selectivity.