Learn More
Waters Corp Waters XBridge BEH Phenyl XP Column, 130Å, 2.5 μm, 4.6 mm × 150 mm
Catalog No. 50466428 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The XBridge BEH Phenyl XP Column provides complementary selectivity to C18 stationary phases, ideal for polyaromatic compounds. It features the most stable Phenyl sorbent, ensuring ultra-low column bleed, long lifetimes, and excellent peak shape. Its ultra-low dispersion hardware enables exceptional HPLC separation and seamless UPLC transition, offering flexibility across diverse pH and temperatures.
  • Offers complementary selectivity to C18 stationary phases, particularly for polyaromatic compounds.
  • Utilizes the most chemically stable Phenyl sorbent for ultra-low column bleed and extended column lifetimes.
  • Delivers excellent peak shape and exceptional separation performance in ultra-low dispersion hardware.
  • Facilitates seamless transition from HPLC to UPLC applications.
  • Provides flexibility across a wide range of mobile phase pH and temperatures.
Catalog No. 50-466-428 Supplier Waters Corp Supplier No. 186006720
The XBridge BEH Phenyl XP Column provides complementary selectivity to C18 stationary phases, ideal for polyaromatic compounds. It features the most stable Phenyl sorbent, ensuring ultra-low column bleed, long lifetimes, and excellent peak shape. Its ultra-low dispersion hardware enables exceptional HPLC separation and seamless UPLC transition, offering flexibility across diverse pH and temperatures.
  • Offers complementary selectivity to C18 stationary phases, particularly for polyaromatic compounds.
  • Utilizes the most chemically stable Phenyl sorbent for ultra-low column bleed and extended column lifetimes.
  • Delivers excellent peak shape and exceptional separation performance in ultra-low dispersion hardware.
  • Facilitates seamless transition from HPLC to UPLC applications.
  • Provides flexibility across a wide range of mobile phase pH and temperatures.