Learn More
Waters Corp XBridge BEH Phenyl Column, 130Å, 3.5 μm, 4.6 mm X 250 mm
Catalog No. 502392164 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The columns offer complementary selectivity compared to C18 stationary phases, particularly for polyaromatic compounds. They are noted for their chemical stability, providing ultra-low column bleed, extended column lifetimes, and excellent peak shape. The columns allow for robust method development due to their distinctive bonding and end-capping technology, which enables operation across a wide pH range and temperature conditions.
  • Chemically stable sorbent
  • Ultra-low column bleed
  • Excellent peak shape
  • Complementary selectivity to C18 stationary phases
  • Effective for analytes with an aromatic ring
  • Extraordinary low and high pH stability
  • Allows for orthogonal techniques in challenging separations
  • Compatible with conventional techniques and UPLC technology
Catalog No. 50-239-2164 Supplier Waters Corp Supplier No. 186003965RF
The columns offer complementary selectivity compared to C18 stationary phases, particularly for polyaromatic compounds. They are noted for their chemical stability, providing ultra-low column bleed, extended column lifetimes, and excellent peak shape. The columns allow for robust method development due to their distinctive bonding and end-capping technology, which enables operation across a wide pH range and temperature conditions.
  • Chemically stable sorbent
  • Ultra-low column bleed
  • Excellent peak shape
  • Complementary selectivity to C18 stationary phases
  • Effective for analytes with an aromatic ring
  • Extraordinary low and high pH stability
  • Allows for orthogonal techniques in challenging separations
  • Compatible with conventional techniques and UPLC technology