Learn More
Waters Corp XBridge BEH Phenyl Column, 130 Å, 3.5 μm, 4.6 mm, 100 mm
Catalog No. 502391318 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
XBridge BEH Phenyl columns offer complementary selectivity to C18 stationary phases, ideal for polyaromatic compounds. Featuring the most chemically stable Phenyl sorbent, this product ensures ultra-low column bleed, long lifetimes, and excellent peak shape. An electronic eConnect tag enables automatic identification, tracking, and digital tracing of column use history. Full advantages are realized with compatible systems running Empower software.
  • Complementary selectivity to C18 stationary phases, especially for polyaromatic compounds
  • Most chemically stable commercially available Phenyl sorbent
  • Ultra-low column bleed
  • Long column lifetimes
  • Excellent peak shape
  • Electronic eConnect column tag for automatic identification, tracking, and digital tracing of column use history
  • Enhanced advantages when used with compatible systems running Empower software
Catalog No. 50-239-1318 Supplier Waters Corp Supplier No. 186003334RF
Add to Cart
Add to Cart
XBridge BEH Phenyl columns offer complementary selectivity to C18 stationary phases, ideal for polyaromatic compounds. Featuring the most chemically stable Phenyl sorbent, this product ensures ultra-low column bleed, long lifetimes, and excellent peak shape. An electronic eConnect tag enables automatic identification, tracking, and digital tracing of column use history. Full advantages are realized with compatible systems running Empower software.
  • Complementary selectivity to C18 stationary phases, especially for polyaromatic compounds
  • Most chemically stable commercially available Phenyl sorbent
  • Ultra-low column bleed
  • Long column lifetimes
  • Excellent peak shape
  • Electronic eConnect column tag for automatic identification, tracking, and digital tracing of column use history
  • Enhanced advantages when used with compatible systems running Empower software