Learn More
Waters Corp XBridge Preparative Column, BEH Phenyl, 10 × 100 mm, 5 μm
Supplier:  Waters Corp 186008177
Encompass_Preferred
This XBridge OBD Preparative Column is designed for purification in preparative liquid chromatography (Prep LC). Featuring BEH Phenyl chemistry and 5 μm particle size, this 10 × 100 mm OBD column ensures uniform, optimum packed bed density for consistent performance.
  • Direct scalability from analytical to Prep
  • Excellent column lifetime
  • Consistent column-to-column performance
  • Good resolution, peak shape, and mass loading
  • Universal column choice for a wide variety of compounds
  • Exceptional peak shape for basic analytes at elevated pH
  • Stability across a wide pH range and at high temperatures
Catalog No. 50-239-2666